Please login to your online account to display your discounted pricing

Brucine sulfate hydrate, 98+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC218171000

 View more versions of this product

Catalog No. AC218171000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals or Fine Crystalline Powder
Formula Weight 443.5
Quantity 100g
Color White to slightly beige
Specific Rotation Condition −24° (22°C c=1,H2O)
Assay 98% min. (on dry substance)
Infrared Spectrum Authentic
Residue after Ignition 0.1% max.
CAS 652154-10-4
Assay Percent Range 98+%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Harmful to aquatic life with long lasting effects.
Fatal if swallowed.
MDL Number MFCD00150158
Water 13% max.
Packaging Glass bottle
Sensitivity (to nitrate) Passes Test
Insoluble in Water Passes Test
Chemical Name or Material Brucine sulfate hydrate
Molecular Formula 1/2H2SO4·xH2O

Specifications

Chemical Name or Material Brucine sulfate hydrate
CAS 652154-10-4
Assay 98% min. (on dry substance)
Assay Percent Range 98+%
Molecular Formula 1/2H2SO4·xH2O
Formula Weight 443.5
MDL Number MFCD00150158
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Harmful to aquatic life with long lasting effects.
Fatal if swallowed.

SDS & Safety Documents