missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brucine sulfate hydrate, 98+%, ACROS Organics™

Chemical Identifiers

CAS 652154-10-4
Assay Percent Range 98+%
Molecular Formula 1/2H2SO4·xH2O
Formula Weight 443.5
MDL Number MFCD00150158
Chemical Name or Material Brucine sulfate hydrate
Assay 98% min. (on dry substance)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Qty  
AC218171000 SDS Thermo Fisher Scientific
AC218171000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC218170250 SDS Thermo Fisher Scientific
218170250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

652154-10-4
1/2H2SO4·xH2O
MFCD00150158
98% min. (on dry substance)
98+%
443.5
Brucine sulfate hydrate

Specifications

652154-10-4
Passes Test
(to nitrate) Passes Test
Crystals or Fine Crystalline Powder
1/2H2SO4·xH2O
443.5
−24° (22°C c=1,H2O)
100g
98% min. (on dry substance)
Authentic
Glass bottle
0.1% max.
98+%
13% max.
MFCD00150158
White to slightly beige
Brucine sulfate hydrate

Safety and Handling

GHS H Statement:
Fatal if inhaled.
Harmful to aquatic life with long lasting effects.
Fatal if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger