Please login to your online account to display your discounted pricing

Alfa Aesar™ Brucine sulfate heptahydrate, ACS

Resolving agent

Chemical Identifiers

Chemical Name or Material Brucine sulfate heptahydrate
CAS 60583-39-3
Purity Grade ACS Grade
Formula Weight 1013.15 (887.03 Anhydrous)
MDL Number MFCD00150159
UN Number UN2811
Merck Index 14,1455
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3624914

 
alfa aesar™
3624914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1013.15 (887.03 Anhydrous)
Chemical Name or Material Brucine sulfate heptahydrate
TSCA TSCA
Purity Grade ACS Grade
Physical Form Powder
MDL Number MFCD00150159
CAS 60583-39-3
UN Number UN2811
Merck Index 14,1455
Melting Point 180°C (dec.)
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.