missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brucine dihydrate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 163501000

 View more versions of this product

Catalog No. AC163501000



Specifications

Specifications

Brucine dihydrate
6.0% to 9.5% (105°C, 3 hrs)
-123
174°C to 179°C
99%
99%
MFCD00149384
10,11-dimethoxystrychnine
VWLBJWIPYIYRBM-FIMIILAWSA-N
(4aR,5aS,8aR,13aS,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;dihydrate
69897601
Powder or Crystals
Authentic
Glass bottle
White to Beige
100g
145428-94-0
C23H26N2O4·2H2O
14, 1455
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform and benzene,slightly soluble in ether,glycerol and,ethyl acetate
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC.O.O
430.5
430.5
99%

Chemical Identifiers

145428-94-0
430.5
VWLBJWIPYIYRBM-FIMIILAWSA-N
69897601
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC.O.O
C23H26N2O4·2H2O
MFCD00149384
10,11-dimethoxystrychnine
(4aR,5aS,8aR,13aS,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents