missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brucine dihydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1625109

 View more versions of this product

Catalog No. AAA1625109



Description

Description

Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resolving agentBrucine dihydrate is used as a resolving agent for chiral acids.

Solubility
Soluble in methanol. Partially soluble Miscible with water.

Notes
Incompatible with strong oxidizing agents, bases and strong reducing agents.
Specifications

Specifications

Brucine dihydrate
176°C to 178°C
MFCD00149384
UN1570
YXTWCYYDGRBUDE-UHFFFAOYSA-K
tripotassium;2-hydroxy-2-oxoacetate;iron;oxalate;oxalic acid
131675837
98%
5892-11-5
C6H3FeK3O12
10 g
Soluble in methanol. Partially soluble Miscible with water.
C(=O)(C(=O)O)O.C(=O)(C(=O)[O-])O.C(=O)(C(=O)[O-])[O-].[K+].[K+].[K+].[Fe]
440.22
430.50 (394.47 Anhydrous)

Chemical Identifiers

5892-11-5
MFCD00149384
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c

H300+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BRUCINE

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only