missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brucine, 99%, anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC278725000

 View more versions of this product

Catalog No. AC278725000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Brucine
1% max. (105°C, 3 hrs)
−123° (20°C c=2.5,CCl3)
0.1% max.
174.0°C to 179.0°C
Anhydrous
99%
MFCD00005942
15, 1464
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, chloroform and benzene, slightly soluble in ether, glycerol and, ethyl acetate
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC
394.46
394.46
99%
Authentic
Glass bottle
− 123.00
White to Beige
500g
357-57-3
C23H26N2O4
27, IV, 7876
brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy
RRKTZKIUPZVBMF-VLTSECPOSA-N
(4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
51413923
Crystalline Powder

Chemical Identifiers

357-57-3
394.46
RRKTZKIUPZVBMF-VLTSECPOSA-N
51413923
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC
C23H26N2O4
MFCD00005942
brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy
(4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal if swallowed.
Harmful to aquatic life with long lasting effects.
Fatal if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-614-7

SDS Documents