Learn More
Brucine, 99%, anhydrous, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 278720500
Specifications
Brucine | |
1% max. (105°C, 3 hrs) | |
− 123.00 | |
White to Beige | |
50g | |
357-57-3 | |
C23H26N2O4 | |
27, IV, 7876 | |
brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy | |
RRKTZKIUPZVBMF-VLTSECPOSA-N | |
(4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one | |
51413923 | |
Crystalline Powder |
Authentic | |
Glass bottle | |
0.1% max. | |
174.0°C to 179.0°C | |
Anhydrous | |
99% | |
MFCD00005942 | |
15, 1464 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, chloroform and benzene, slightly soluble in ether, glycerol and, ethyl acetate | |
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC | |
394.46 | |
394.46 | |
99% |
Chemical Identifiers
357-57-3 | |
394.46 | |
RRKTZKIUPZVBMF-VLTSECPOSA-N | |
51413923 | |
COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC |
C23H26N2O4 | |
MFCD00005942 | |
brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy | |
(4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one |
Safety and Handling
GHS H Statement:
Fatal if swallowed.
Harmful to aquatic life with long lasting effects.
Fatal if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 206-614-7