Allied National Brother™ Laminated Adhesive Tape for PT-550 Labeling System
Manufacturer:  Allied National TZB51
 View more versions of this product
Catalog No. 11-877-31
Description
Description
This tape features our exclusive laminated tape process and our standard adhesive. Perfect for everyday applications. These labels stay on under normal indoor and outdoor use, including hot and cold environments. Best suited for smooth flat surfaces.
  • Labeling tape for your Brother labelling system.
  • Black print on white tape.
  • 1-inch width.
  • Compatible with PT 330, 350, 520, 540, 580C, 2500PC, 2200, 2400, 530, 550, 9200PC, and 9400 models.
Specifications
Specifications
Black/Fluorescent Orange
Tape
Tape
1 in.