Please login to your online account to display your discounted pricing

Bropirimine, Tocris™

Immunomodulatory and antitumor compound

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Bropirimine
Assay Percent Range >99%
Molecular Formula C10H8BrN3O
Formula Weight 266.09
Synonym 2-Amino-5-bromo-6-phenyl-4(3H)-pyrimidone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

431610

 
r & d systems
431610
10mg Each for $164.61

431650

 
r & d systems
431650
50mg Each for $682.67
Description & Specifications

Specifications

Chemical Name or Material Bropirimine
CAS 56741-95-8
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >99%
Molecular Formula C10H8BrN3O
Formula Weight 266.09
Synonym 2-Amino-5-bromo-6-phenyl-4(3H)-pyrimidone
Quantity 10mg
Safety and Handling

Desiccate at Room Temperature