missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Brompheniramine maleate

An antihistamine and also an inhibitor of serotonin reuptake

$121.68 - $404.04

Chemical Identifiers

CAS 980-71-2
Molecular Formula C20H23BrN2O4
Molecular Weight (g/mol) 435.318
MDL Number MFCD00865691
InChI Key SRGKFVAASLQVBO-BTJKTKAUSA-N
Synonym brompheniramine maleate, brompheniramine hydrogen maleate, dimetane, rolabromophen, dimotane, drauxin, gammistin, nagemid, spentane, allent
PubChem CID 5281067
ChEBI CHEBI:3184
IUPAC Name 3-(4-bromophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine;(Z)-but-2-enedioic acid
SMILES CN(C)CCC(C1=CC=C(C=C1)Br)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63922MB View Documents Alfa Aesar™
J63922MB
25mg Each for $121.68
 
AAJ63922MC View Documents Alfa Aesar™
J63922MC
100mg Each for $404.04
 
Specifications

Chemical Identifiers

980-71-2
435.318
SRGKFVAASLQVBO-BTJKTKAUSA-N
5281067
3-(4-bromophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine;(Z)-but-2-enedioic acid
C20H23BrN2O4
MFCD00865691
brompheniramine maleate, brompheniramine hydrogen maleate, dimetane, rolabromophen, dimotane, drauxin, gammistin, nagemid, spentane, allent
CHEBI:3184
CN(C)CCC(C1=CC=C(C=C1)Br)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O

Specifications

White
980-71-2
MFCD00865691
brompheniramine maleate, brompheniramine hydrogen maleate, dimetane, rolabromophen, dimotane, drauxin, gammistin, nagemid, spentane, allent
CN(C)CCC(C1=CC=C(C=C1)Br)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O
435.318
CHEBI:3184
Powder
25mg
C20H23BrN2O4
14,1443
SRGKFVAASLQVBO-BTJKTKAUSA-N
3-(4-bromophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine;(Z)-but-2-enedioic acid
5281067
319.24
Brompheniramine maleate
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA