Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromotris(triphenylphosphine)rhodium(I), 99.95% (metals basis), Rh 10.1% min

Hydrogenation

Chemical Identifiers

Chemical Name or Material Bromotris(triphenylphosphine)rhodium(I)
CAS 14973-89-8
Formula Weight 969.69
MDL Number MFCD00009828
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3982101

  SDS
alfa aesar™
3982101
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3982103

  SDS
alfa aesar™
3982103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3982106

  SDS
alfa aesar™
3982106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 969.69
Chemical Name or Material Bromotris(triphenylphosphine)rhodium(I)
Physical Form Crystalline
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00009828
CAS 14973-89-8
Concentration Rh 10.1% min.
Quantity 0.25g
Solubility Information Soluble in chlorinated solvents