Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromotris(dimethylamino)phosphonium hexafluorophosphate, 98+%

Chemical Identifiers

Chemical Name or Material Bromotris(dimethylamino)phosphonium hexafluorophosphate
CAS 50296-37-2
Molecular Formula C6H18BrF6N3P2
Formula Weight 388.07
MDL Number MFCD00191864
UN Number UN1759
Synonym BroP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2670803

 
alfa aesar™
H2670803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2670806

 
alfa aesar™
H2670806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 388.07
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Chemical Name or Material Bromotris(dimethylamino)phosphonium hexafluorophosphate
Assay ≥98%
Molecular Formula C6H18BrF6N3P2
MDL Number MFCD00191864
CAS 50296-37-2
UN Number UN1759
Melting Point >300°C
Synonym BroP
Quantity 1g
Health Hazard 2 GHS H Statement
H350-H314-H318
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H314-H318
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C