Please login to your online account to display your discounted pricing

Bromotriphenylethylene, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  209070050

 View more versions of this product

Catalog No. AC209070050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 335.23
Quantity 5g
Color Light yellow
EINECS Number 216-531-8
Melting Point 114° to 118°C
RTECS Number KU8850000
Beilstein 05, 722
Chemical Name or Material Bromotriphenylethylene
Name Note 98%
Molecular Formula C20 H15 Br
Linear Formula (C6H5)2C=C(C6H5)Br
CAS 1607-57-4
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00000135
Packaging Glass bottle

Specifications

Chemical Name or Material Bromotriphenylethylene
Name Note 98%
CAS 1607-57-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C20 H15 Br
Linear Formula (C6H5)2C=C(C6H5)Br
Formula Weight 335.23
MDL Number MFCD00000135