missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromotriphenylethylene, 98%, ACROS Organics™

Chemical Identifiers

CAS 1607-57-4
Assay Percent Range 98%
Molecular Formula C20 H15 Br
Linear Formula (C6H5)2C=C(C6H5)Br
Formula Weight 335.23
MDL Number MFCD00000135
Chemical Name or Material Bromotriphenylethylene, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC209070050 SDS Thermo Fisher Scientific
209070050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1607-57-4
C20 H15 Br
335.23
Bromotriphenylethylene, 98%
98%
(C6H5)2C=C(C6H5)Br
MFCD00000135
≥97.5%

Specifications

335.23
5g
114° to 118°C
Authentic
1607-57-4
MFCD00000135
Glass bottle
≥97.5%
Crystalline Powder
Light yellow
05, 722
(C6H5)2C=C(C6H5)Br
C20 H15 Br
98%
Bromotriphenylethylene, 98%

Safety and Handling

EINECSNumber : 216-531-8

RTECSNumber : KU8850000