missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromotrimethylsilane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 186650050

 View more versions of this product

Catalog No. AC186650050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Silylation reagent
Specifications

Specifications

Bromotrimethylsilane
-43.0°C
1.1600g/mL
32°C
97.5% min. (Argentometry)
C3H9BrSi
(CH3)3SiBr
09,73; 10,59; 12,79; 15,51; 16,50
trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
IYYIVELXUANFED-UHFFFAOYSA-N
bromo(trimethyl)silane
76113
98%
2857-97-8
Colorless to Yellow
79.0°C
Authentic
Glass bottle
1.4200 to 1.4250
5 g
1.16
Solubility in water: decomposes.
C[Si](C)(C)Br
153.1
153.1
Liquid

Chemical Identifiers

2857-97-8
153.1
trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
bromo(trimethyl)silane
C3H9BrSi
IYYIVELXUANFED-UHFFFAOYSA-N
76113
C[Si](C)(C)Br
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 220-672-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only