missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromotrimethylsilane, 97%, stab. with copper powder or silver wire

Manufacturer:  Alfa Aesar™ A1533430

 View more versions of this product

Catalog No. AAA1533430



Description

Description

Applications
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.

Solubility
Miscible with carbon tetrachloride, chloroform, dichloroethane, acetonitrile, toluene, dichloromethane and hexane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids, water and strong oxidizing agents.
Specifications

Specifications

Bromotrimethylsilane
2857-97-8
(CH3)3SiBr
UN2924
trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
C[Si](C)(C)Br
153.094
153.09
Acrid
-44°C
−3°C (26°F)
stabilized with copper powder or silver wire
C3H9BrSi
MFCD00000048
1731135
IYYIVELXUANFED-UHFFFAOYSA-N
bromo(trimethyl)silane
76113
97%
78°C to 80°C
250g

Chemical Identifiers

2857-97-8
153.094
IYYIVELXUANFED-UHFFFAOYSA-N
76113
C[Si](C)(C)Br
C3H9BrSi
MFCD00000048
trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
bromo(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents