missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromotrichloromethane 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107492500

 View more versions of this product

Catalog No. AC107492500


 

Specifications Description & Specifications

Specifications

Liquid
≥98.5%
Clear colorless to slightly yellow
01,80; 02,42; 07,40
99%
CBrCl3
Bromotrichloromethane
BrCCl3
Authentic
198.27
250mL
01, 67
1.5055 to 1.5075 (20°C, 589nm)
99%
MFCD00000783
75-62-7
Glass bottle

Chemical Identifiers

75-62-7
CBrCl3
198.27
Bromotrichloromethane, 99%
99%
BrCCl3
MFCD00000783
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-886-0

RTECSNumber : PA5400000

TSCA : TSCA