missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Bromothymol Blue Solution

Manufacturer:  Fisherbrand™ S718891

 View more versions of this product

Catalog No. S718891



Description

Description

CO2/pH indicator

Specifications

Specifications

Bromothymol Blue Solution
C27H27Br2NaO5S
MFCD00077263,MFCD00077263,MFCD00077263
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
646.37
646.37
Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Blue
34722-90-2
C27H27Br2O5SNa
NMKFVGALBGZKGW-FKWCIMQXSA-M
sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
102183223
0.04% Aqueous Solution
pH 6.0-7.6 (yellow-blue)
100mL

Chemical Identifiers

34722-90-2
646.37
NMKFVGALBGZKGW-FKWCIMQXSA-M
sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
C27H27Br2NaO5S
MFCD00077263,MFCD00077263,MFCD00077263
102183223
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS
Documents