Please login to your online account to display your discounted pricing

Bromothymol Blue Solution

$6.65 - $6.65

Chemical Identifiers

Chemical Name or Material Bromothymol Blue Solution
CAS 34722-90-2
Linear Formula C27H27Br2O5SNa
Formula Weight 646.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S718891

 
fisher scientific
S718891
0.04% Aqueous Solution 100mL Each for $6.65
Description & Specifications

Specifications

Formula Weight 646.37
Quantity 100mL
Color Blue
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
pH Color Changes pH 6.0-7.6 (yellow-blue)
Chemical Name or Material Bromothymol Blue Solution
Shelf Life 12 months
Linear Formula C27H27Br2O5SNa
CAS 34722-90-2
ChemAlert Storage Symbol Gray
Packaging 0.04% Aqueous Solution

CO2/pH indicator

Safety and Handling
 

12 months