Please login to your online account to display your discounted pricing

Bromothymol Blue Sodium Salt, ACS Grade, LabChem™

$159.91 - $159.91

Chemical Identifiers

Chemical Name or Material Bromothymol Blue Sodium Salt
CAS 34722-90-2
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C27H27Br2O5S·Na
Formula Weight 646.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC120407

 
lab chem
LC120407
Amber Glass 10g Each for $159.91
Description & Specifications

Specifications

Formula Weight 646.37
Chemical Name or Material Bromothymol Blue Sodium Salt
Quantity 10g
Purity Grade ACS Grade
Physical Form Powder
Linear Formula C27H27Br2O5S·Na
CAS 34722-90-2
Identification Passes Test
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; Hydrogen bromide
Color Gray-orange
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature