Please login to your online account to display your discounted pricing

Bromothymol Blue-Phenol Red, Mixed Indicator, Certified, LabChem™

$32.89 - $32.89

Chemical Identifiers

Chemical Name or Material Bromothymol Blue-Phenol Red
CAS 7732-18-5
CAS Max % 99.95
CAS 34722-90-2
CAS Max % 0.03
CAS 34487-61-1
CAS Max % 0.02
Name Note Mixed Indicator
Purity Grade Certified Grade
Linear Formula C27H27Br2O5S·Na
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC120807

 
lab chem
LC120807
Poly Bottle 125mL Each for $32.89
Description & Specifications

Specifications

Formula Weight 646.37
Purity Grade Certified Grade
Chemical Name or Material Bromothymol Blue-Phenol Red
Specific Gravity 1g/mL
CAS Max % 0.02
Packaging Poly Bottle
CAS Max % 99.95
CAS Max % 0.03
Density 1g/mL
Quantity 125mL
Linear Formula C27H27Br2O5S·Na
Solubility Soluble in water
Identification Passes Test
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Recommended Storage Room Temperature
CAS 7732-18-5
CAS 34722-90-2
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Color Dark green
CAS 34487-61-1
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Name Note Mixed Indicator
Physical Form Liquid
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature