missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromothymol Blue, ACS

Manufacturer:  Alfa Aesar™ 3263806

 View more versions of this product

Catalog No. AA3263806



Specifications

Specifications

Bromothymol Blue
ACS Grade
MFCD00005872
14,1445
NUHCTOLBWMJMLX-UHFFFAOYSA-N
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
6450
624.39
200°C to 202°C
5g
76-59-5
C27H28Br2O5S
373934
Soluble in alcohol and aqueous solutions of alkali; sparingly soluble in water
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
624.384
CHEBI:86155
Crystalline
Brown to Red

Chemical Identifiers

76-59-5
624.384
NUHCTOLBWMJMLX-UHFFFAOYSA-N
CHEBI:86155
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
C27H28Br2O5S
MFCD00005872
6450
2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
Safety and Handling

Safety and Handling

RTECSNumber : SJ450000

TSCA : TSCA

SDS Documents