Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromosuccinic acid, 96%

$57.57 - $310.39

Chemical Identifiers

Chemical Name or Material Bromosuccinic acid
CAS 923-06-8
Molecular Formula C4H5BrO4
Formula Weight 196.99
MDL Number MFCD00004213
Beilstein 1723556
Merck Index 14,1437
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2282906

  SDS
alfa aesar™
B2282906
5g Each for $57.57

AAB2282922

  SDS
alfa aesar™
B2282922
100g Each for $310.39
Description & Specifications

Specifications

Formula Weight 196.99
Quantity 5g
CAS 923-06-8
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00004213
Merck Index 14,1437
Melting Point 164° to 166°C (dec.)
Assay 96%
Chemical Name or Material Bromosuccinic acid
Beilstein 1723556
Molecular Formula C4H5BrO4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning

Applications
Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.

Solubility
Insoluble in ether.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.