Learn More
Bromosuccinic acid, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2282922
Description
Bromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromosuccinic acid is an important precursor in many chemicals used in the food, chemical and pharmaceutical fields. It is also used as a reagent to prepare L-Hydrazinosuccinic acid.
Solubility
Insoluble in ether.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications
Bromosuccinic acid | |
164°C to 166°C (decomposition) | |
MFCD00004213 | |
1723556 | |
bromosuccinic acid, 2-bromosuccinic acid, monobromosuccinic acid, butanedioic acid, bromo, +--bromosuccinic acid, dl-bromosuccinic acid, rs-bromosuccinic acid, succinic acid, bromo, bromobutanedioic acid | |
QQWGVQWAEANRTK-UHFFFAOYNA-N | |
2-bromobutanedioic acid | |
73557 | |
196.99 |
923-06-8 | |
C4H5BrO4 | |
100 g | |
14,1437 | |
Insoluble in ether. | |
OC(=O)CC(Br)C(O)=O | |
196.98 | |
CHEBI:73712 | |
96% |
Chemical Identifiers
923-06-8 | |
196.98 | |
QQWGVQWAEANRTK-UHFFFAOYNA-N | |
73557 | |
2-bromobutanedioic acid |
C4H5BrO4 | |
MFCD00004213 | |
bromosuccinic acid, 2-bromosuccinic acid, monobromosuccinic acid, butanedioic acid, bromo, +--bromosuccinic acid, dl-bromosuccinic acid, rs-bromosuccinic acid, succinic acid, bromo, bromobutanedioic acid | |
CHEBI:73712 | |
OC(=O)CC(Br)C(O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 213-087-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only