missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

β-Bromostyrene, 97%, mixture of cis/trans isomers, ACROS Organics™

Manufacturer:  Acros Organics AC169291000

 View more versions of this product

Catalog No. AC169291000


 

Specifications

Specifications

β-Bromostyrene
103-64-0
Authentic
Glass bottle
1.4200g/mL
110.0°C to 112.0°C (20.0 mmHg)
79°C
Yellow through Brown
C8H7Br
MFCD00000185
YMOONIIMQBGTDU-VOTSOKGWSA-N
[(E)-2-bromoethenyl]benzene
5314126
Liquid
Mixture of cis and trans
96.0
100.0
1.6060 to 1.6090
7.0°C
100mL
97%
97%
C6H5CH=CHBr
05, 477
C1=CC=C(C=C1)C=CBr
183.048
183.05

Chemical Identifiers

103-64-0
183.048
YMOONIIMQBGTDU-VOTSOKGWSA-N
C8H7Br
MFCD00000185
5314126
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents