missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
beta-Bromostyrene, 97%, mixture of cis/trans isomers, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 169290050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
β-Bromostyrene | |
103-64-0 | |
100.0 | |
110.0°C to 112.0°C (20.0 mmHg) | |
Authentic | |
Glass bottle | |
1.6060 to 1.6090 | |
MFCD00000185 | |
05, 477 | |
2-bromovinyl benzene, beta-bromostyrene, styryl bromide, 2-bromoethenyl benzene, bromostyrolene, 1-bromo-2-phenylethene, bromostyrol, bromstyrole, hyacinth base, 1-bromo-2-phenylethylene | |
YMOONIIMQBGTDU-VOTSOKGWSA-N | |
[(E)-2-bromoethenyl]benzene | |
5314126 | |
97% |
Mixture of cis and trans | |
96.0 | |
1.4200g/mL | |
79°C | |
96% min. (sum of isomers) (GC) | |
C8H7Br | |
C6H5CH=CHBr | |
5mL | |
1.42 | |
Solubility in water: .. Other solubilities: soluble in alcohol | |
C1=CC=C(C=C1)C=CBr | |
183.05 | |
183.05 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 203-131-3
RUO â Research Use Only