Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromopolystyrene, ca 1.0-2.0 mmol/g

Chemical Identifiers

Chemical Name or Material Bromopolystyrene
MDL Number MFCD00146196
Synonym Polystyrene, brominated
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0794106

  SDS
alfa aesar™
L0794106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0794114

  SDS
alfa aesar™
L0794114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Bromopolystyrene
Concentration ≈1.0 to 2.0mmol/g
Quantity 5g
Synonym Polystyrene, brominated
MDL Number MFCD00146196