Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromopolystyrene, ca 1.0-2.0 mmol/g


Quantity 5g

Manufacturer: alfa aesar™  L0794106

 View more versions of this product

Catalog No. AAL0794106

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Bromopolystyrene
MDL Number MFCD00146196
Synonym Polystyrene, brominated
Concentration ≈1.0 to 2.0mmol/g
Quantity 5g

Specifications

Chemical Name or Material Bromopolystyrene
MDL Number MFCD00146196
Synonym Polystyrene, brominated