Please login to your online account to display your discounted pricing

Bromophenol Blue TS, 0.1% (w/v) in dilute Alcohol, Ricca Chemical


Quantity 120mL
Packaging Poly natural

Manufacturer: ricca chemical company  RI13574

 View more versions of this product

Catalog No. 1357-4

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear dark red
Quantity 120mL
Concentration or Composition Notes 0.1% (w/v)
Solvent or Matrix Dilute Alcohol
Chemical Name or Material Bromophenol Blue
CAS Max % 0.11
Name Note Test Solution
CAS 7732-18-5
CAS Min % 54.0
CAS Max % 57.0
CAS 64-17-5
CAS Min % 43.51
CAS Max % 45.28
CAS Min % 0.11
CAS 7732-18-5 (Water, Deionized)
Packaging Poly natural
CAS 62625-28-9

Specifications

Chemical Name or Material Bromophenol Blue
Name Note Test Solution
CAS 7732-18-5
CAS Min % 54.0
CAS Max % 57.0
CAS 64-17-5
CAS Min % 43.51
CAS Max % 45.28
CAS 62625-28-9
CAS Min % 0.11
CAS Max % 0.11
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid