missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromophenol Blue TS, 0.1% (w/v) in dilute Alcohol, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI135716

 View more versions of this product

Catalog No. 1357-16

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Bromophenol Blue
7732-18-5,64-17-5,62625-28-9,7732-18-5 (Water, Deionized)
57.0,45.28,0.11
Poly Natural
Dilute Alcohol
500mL
Test Solution
54.0,43.51,0.11
Liquid
0.1% (w/v)
Clear dark red

Chemical Identifiers

7732-18-5,64-17-5,62625-28-9,7732-18-5 (Water, Deionized)
57.0,45.28,0.11
Bromophenol Blue, Test Solution
54.0,43.51,0.11
Liquid