Please login to your online account to display your discounted pricing

Bromophenol Blue TS, 0.1% (w/v) in dilute Alcohol, Ricca Chemical

$144.30 - $144.30

Chemical Identifiers

Chemical Name or Material Bromophenol Blue
Name Note Test Solution
CAS 7732-18-5
CAS Min % 54.0
CAS Max % 57.0
CAS 64-17-5
CAS Min % 43.51
CAS Max % 45.28
CAS 62625-28-9
CAS Min % 0.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

1357-4

  SDS
ricca chemical company
RI13574
Poly natural 120mL Each for $144.30

1357-16

 
ricca chemical company
RI135716
Poly Natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear dark red
Quantity 120mL
Concentration or Composition Notes 0.1% (w/v)
Solvent or Matrix Dilute Alcohol
Chemical Name or Material Bromophenol Blue
CAS Max % 0.11
Name Note Test Solution
CAS 7732-18-5
CAS Min % 54.0
CAS Max % 57.0
CAS 64-17-5
CAS Min % 43.51
CAS Max % 45.28
CAS Min % 0.11
CAS 7732-18-5 (Water, Deionized)
Packaging Poly natural
CAS 62625-28-9