Please login to your online account to display your discounted pricing

Bromophenol Blue Sodium Salt Molecular Biology MP Biomedicals

$65.38 - $226.22

Chemical Identifiers

Chemical Name or Material Bromothymol Blue sodium salt
Name Note Chemistry
CAS 34722-90-2
Purity Grade Molecular Biology Grade
Physical Form Powder
Molecular Formula C27H27BrNaO5S
Formula Weight 646.39
Synonym 3, 3-dibromothymolsulfonephthalein sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19399005

 
mp biomedicals
19399005
5g Each for $65.38

ICN19399025

 
mp biomedicals
19399025
25g Each for $226.22
Description & Specifications

Specifications

Color Olive-green
Purity Grade Molecular Biology Grade
Physical Form Powder
Formula Weight 646.39
Quantity 5g
EINECS Number 252-169-7
Chemical Name or Material Bromothymol Blue sodium salt
Name Note Chemistry
Synonym 3, 3-dibromothymolsulfonephthalein sodium salt
Molecular Formula C27H27BrNaO5S
CAS 34722-90-2
Recommended Storage Store at Room Temperature (15° to 30°C)
Safety and Handling

Store at Room Temperature (15° to 30°C)