Please login to your online account to display your discounted pricing

Bromophenol Blue Sodium Salt Molecular Biology MP Biomedicals


Quantity 25g

Manufacturer: mp biomedicals  19399025

 View more versions of this product

Catalog No. ICN19399025

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Physical Form Powder
Formula Weight 646.39
Color Olive-green
Quantity 25g
EINECS Number 252-169-7
Name Note Chemistry
Molecular Formula C27H27BrNaO5S
Chemical Name or Material Bromothymol Blue sodium salt
CAS 34722-90-2
Synonym 3, 3-dibromothymolsulfonephthalein sodium salt
Recommended Storage Store at Room Temperature (15° to 30°C)

Specifications

Chemical Name or Material Bromothymol Blue sodium salt
Name Note Chemistry
CAS 34722-90-2
Purity Grade Molecular Biology Grade
Physical Form Powder
Molecular Formula C27H27BrNaO5S
Formula Weight 646.39
Synonym 3, 3-dibromothymolsulfonephthalein sodium salt
Accessories

Store at Room Temperature (15° to 30°C)