Please login to your online account to display your discounted pricing

Bromophenol Blue Sodium Salt, ACS Grade, LabChem™

$80.09 - $80.09

Chemical Identifiers

Chemical Name or Material Bromophenol Blue Sodium Salt
CAS 62625-28-9
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C19H9Br4O5S·Na
Formula Weight 691.95
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC120107

 
lab chem
LC120107
Amber Glass 10g Each for $80.09
Description & Specifications

Specifications

Formula Weight 691.95
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; Hydrogen bromide
Physical Form Powder
CAS Max % 100
CAS 62625-28-9
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Recommended Storage Room Temperature
Quantity 10g
Identification Passes Test
Purity Grade ACS Grade
Chemical Name or Material Bromophenol Blue Sodium Salt
Health Hazard 2 GHS H Statement
Product is not hazardous.
Linear Formula C19H9Br4O5S·Na
Color Dark
Packaging Amber Glass
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature