Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromophenol Blue sodium salt, 0.04% w/v aq. soln.

Chemical Identifiers

Chemical Name or Material Bromophenol Blue sodium salt
CAS 34725-61-6
Formula Weight 691.96
MDL Number MFCD00013793
Merck Index 14,1444
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38693AE

  SDS
alfa aesar™
38693AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38693AP

  SDS
alfa aesar™
38693AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38693M0

  SDS
alfa aesar™
38693M0
6 x 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 691.96
Chemical Name or Material Bromophenol Blue sodium salt
Quantity 100mL
TSCA TSCA
Concentration 0.04% w/v aq. soln.
Physical Form Liquid
MDL Number MFCD00013793
CAS 34725-61-6
Merck Index 14,1444
Color Dark red