Please login to your online account to display your discounted pricing

Bromophenol Blue, pure, 0.1% in water, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC403151000

 View more versions of this product

Catalog No. AC403151000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 669.96
Quantity 100mL
Density 1g/mL
Purity Grade Pure
Concentration about 0.1% (by UV)
CAS 115-39-9
Synonym 3', 3'', 5'
Packaging Glass bottle
Chemical Name or Material Bromophenol Blue
Molecular Formula C19H10Br4O5S
CAS 7732-18-5

Specifications

Chemical Name or Material Bromophenol Blue
CAS 115-39-9
CAS 7732-18-5
Purity Grade Pure
Molecular Formula C19H10Br4O5S
Formula Weight 669.96
Synonym 3', 3'', 5'