missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromophenol Blue Indicator, 0.1% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI135332

 View more versions of this product

Catalog No. 135332


 

Specifications Description & Specifications

Specifications

Bromophenol Blue Indicator
98.0,0.1
Liquid
0.1% (w/v)
Clear dark reddish purple
7732-18-5,62625-28-9
100.0,0.1
Poly natural
Aqueous Solution
1L

Chemical Identifiers

7732-18-5,62625-28-9
100.0,0.1
Bromophenol Blue Indicator
98.0,0.1
Liquid