missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromophenol Blue Indicator, 0.04% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI13504

 View more versions of this product

Catalog No. 1350-4


 

Specifications Description & Specifications

Specifications

Liquid
120mL
Aqueous Solution
Bromophenol Blue Indicator
98.0,0.04
Clear dark reddish purple
0.04% (w/v)
Plastic Bottle
7732-18-5,62625-28-9
100.0,0.04

Chemical Identifiers

7732-18-5,62625-28-9
100.0,0.04
Bromophenol Blue Indicator
98.0,0.04
Liquid