Please login to your online account to display your discounted pricing

Bromophenol Blue Indicator, 0.04% (w/v) Aqueous Solution, Ricca Chemical


Quantity 500mL
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RI135016

 View more versions of this product

Catalog No. 1350-16

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear dark reddish purple
Quantity 500mL
Concentration or Composition Notes 0.04% (w/v)
Solvent or Matrix Aqueous Solution
Chemical Name or Material Bromophenol Blue Indicator
CAS Min % 98.0
Packaging Plastic Bottle
CAS Max % 100.0
CAS Min % 0.04
CAS Max % 0.04
CAS 7732-18-5
CAS 62625-28-9

Specifications

Chemical Name or Material Bromophenol Blue Indicator
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 62625-28-9
CAS Min % 0.04
CAS Max % 0.04
Physical Form Liquid