Please login to your online account to display your discounted pricing

Bromophenol Blue Indicator, 0.04% (w/v) Aqueous Solution, Ricca Chemical


Quantity 250mL
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR1350000250A

 View more versions of this product

Catalog No. R1350000250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear dark reddish purple
Quantity 250mL
Chemical Name or Material Bromophenol Blue Indicator
CAS 7732-18-5
CAS Min % 98.0
Packaging Plastic Bottle
CAS Max % 100.0
CAS 62625-28-9
CAS Min % 0.04
CAS Max % 0.04

Specifications

Chemical Name or Material Bromophenol Blue Indicator
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 62625-28-9
CAS Min % 0.04
CAS Max % 0.04
Physical Form Liquid