missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromophenol Blue (Free Acid), Fisher BioReagents

Manufacturer:  Fisher BioReagents BP11525

Catalog No. BP115-25




Description

Description

Glass Bottle; 25g

Bromophenol Blue is commonly used as a tracking dye in electrophoresis.
Specifications

Specifications

Bromophenol Blue
115-39-9
C19H10Br4O5S
15, 1454
UDSAIICHUKSCKT-UHFFFAOYSA-N
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
8272
5% max.
669.96
Glass Bottle
279°C
25g
Free Acid
Vapor Pressure: Negligible
1000g(-1) cm(-1) min.
Pass Test
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
669.96
589 to 593nm
CHEBI:59424
Solid
Solubility in water: Soluble
Violet
SDS
Documents
Product Certifications
Promotions

Promotions