missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromophenol Blue, 0.1% Aqueous, pH 3.0 to 4.6 Yellow to Blue, Certified, LabChem™

Manufacturer:  LabChem LC120307

 View more versions of this product

Catalog No. LC120307



Specifications

Specifications

Bromophenol Blue
62625-28-9,7732-18-5
C19H9Br4NaO5S
Passes Test
C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
691.942
691.95
Certified
Soluble in water
Purple-Red
0.1% Aqueous, pH 3.0 to 4.6 Yellow to Blue
99.9,0.1
C19H9Br4O5S·Na
TWKDJMRHSLWRFI-UHFFFAOYSA-M
sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
3791690
Liquid
Poly Bottle
Hydrogen bromide; Carbon dioxide; Carbon monoxide; Nitrogen oxides
125mL

Chemical Identifiers

62625-28-9
691.942
3791690
C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
C19H9Br4NaO5S
TWKDJMRHSLWRFI-UHFFFAOYSA-M
sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

Documents
Promotions

Promotions