Please login to your online account to display your discounted pricing

β-Bromophenetole 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  107251000

 View more versions of this product

Catalog No. AC107251000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 201.06
Physical Form Liquid After Melting
Quantity 100g
Color Clear colorless
Beilstein 06, 142
EINECS Number 209-634-4
TSCA TSCA
CAS Max % 100.0
Synonym β-Bromophenylethyl ether
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Chemical Name or Material β-Bromophenetole
Refractive Index 1.5550 to 1.5570 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H9BrO
Linear Formula C6H5OCH2CH2Br
MDL Number MFCD00000234
CAS 589-10-6
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material β-Bromophenetole
Name Note 98%
CAS 589-10-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9BrO
Linear Formula C6H5OCH2CH2Br
Formula Weight 201.06
MDL Number MFCD00000234
Synonym β-Bromophenylethyl ether
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents