Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromopentamethylbenzene, 98+%

$44.15 - $125.07

Chemical Identifiers

Chemical Name or Material Bromopentamethylbenzene
CAS 5153-40-2
Molecular Formula C11H15Br
Formula Weight 227.15
MDL Number MFCD00013523
Beilstein 1863555
Synonym Pentamethylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0245106

  SDS
alfa aesar™
L0245106
5g Each for $44.15

AAL0245114

  SDS
alfa aesar™
L0245114
25g Each for $125.07
Description & Specifications

Specifications

Formula Weight 227.15
Chemical Name or Material Bromopentamethylbenzene
MDL Number MFCD00013523
Assay ≥98%
Beilstein 1863555
Synonym Pentamethylbromobenzene
Melting Point 161° to 162°C
Quantity 5g
Molecular Formula C11H15Br
CAS 5153-40-2

Applications
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.