missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bromopentamethylbenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0245106
Description
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Bromopentamethylbenzene | |
161°C to 162°C | |
MFCD00013523 | |
1863555 | |
Insoluble in water. | |
CC1=C(C)C(C)=C(Br)C(C)=C1C | |
227.15 | |
227.15 |
5153-40-2 | |
C11H15Br | |
5 g | |
bromopentamethylbenzene, benzene, bromopentamethyl, pentamethylbromobenzene, 1-bromopentamethylbenzene, 1-bromo-2,3,4,5,6-pentamethyl-benzene, brompentamethylbenzol, maybridge1_002545, ksc273s6f, acmc-1b227, 5-bromo-1,2,3,4,6-pentamethylbenzene | |
XPDQRULPGCFCLX-UHFFFAOYSA-N | |
1-bromo-2,3,4,5,6-pentamethylbenzene | |
78831 | |
≥98% |
Chemical Identifiers
5153-40-2 | |
227.15 | |
XPDQRULPGCFCLX-UHFFFAOYSA-N | |
78831 | |
CC1=C(C)C(C)=C(Br)C(C)=C1C |
C11H15Br | |
MFCD00013523 | |
bromopentamethylbenzene, benzene, bromopentamethyl, pentamethylbromobenzene, 1-bromopentamethylbenzene, 1-bromo-2,3,4,5,6-pentamethyl-benzene, brompentamethylbenzol, maybridge1_002545, ksc273s6f, acmc-1b227, 5-bromo-1,2,3,4,6-pentamethylbenzene | |
1-bromo-2,3,4,5,6-pentamethylbenzene |
Safety and Handling
EINECSNumber : 225-926-4
TSCA : No
RUO – Research Use Only