missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bromopentafluoroacetone, 95%

Manufacturer:  Alfa Aesar™ L1378906

 View more versions of this product

Catalog No. AAL1378906



Specifications

Specifications

Bromopentafluoroacetone
5g
C3BrF5O
UN3265
bromopentafluoroacetone, bromopentafluoro-2-propanone, bromopentafluoroacetone, bromopentafluoropropan-2-one, 1-bromo-1,1,3,3,3-pentafluoroacetone, 3-bromo-1,1,1,3,3-pentafluoroacetone, 1-bromo-1,1,3,3,3-pentafluoro-2-propanone, 2-propanone,1-bromo-1,1,3,3,3-pentafluoro, 1-bromanyl-1,1,3,3,3-pentakis fluoranyl propan-2-one
C(=O)(C(F)(F)F)C(F)(F)Br
226.928
226.93
31°C to 33°C
815-23-6
MFCD00153080
1773590
IYDJQZOFWOSTFO-UHFFFAOYSA-N
1-bromo-1,1,3,3,3-pentafluoropropan-2-one
2736352
95%

Chemical Identifiers

815-23-6
226.928
IYDJQZOFWOSTFO-UHFFFAOYSA-N
2736352
C(=O)(C(F)(F)F)C(F)(F)Br
C3BrF5O
MFCD00153080
bromopentafluoroacetone, bromopentafluoro-2-propanone, bromopentafluoroacetone, bromopentafluoropropan-2-one, 1-bromo-1,1,3,3,3-pentafluoroacetone, 3-bromo-1,1,1,3,3-pentafluoroacetone, 1-bromo-1,1,3,3,3-pentafluoro-2-propanone, 2-propanone,1-bromo-1,1,3,3,3-pentafluoro, 1-bromanyl-1,1,3,3,3-pentakis fluoranyl propan-2-one
1-bromo-1,1,3,3,3-pentafluoropropan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents