Please login to your online account to display your discounted pricing

Bromomethyl methyl ether, 90%, Acros Organics

$46.72 - $46.72

Chemical Identifiers

Chemical Name or Material Bromomethyl methyl ether
Assay Percent Range 90%
Molecular Formula C2H5BrO
Formula Weight 124.97
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171650250

  SDS
thermo fisher scientific
AC171650250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC171650050

  SDS
thermo fisher scientific
AC171650050
Glass bottle 5g Each for $46.72
Description & Specifications

Specifications

Formula Weight 124.97
Quantity 25g
Chemical Name or Material Bromomethyl methyl ether
Assay Percent Range 90%
Molecular Formula C2H5BrO
CAS 13057-17-5
Packaging Glass bottle