Please login to your online account to display your discounted pricing

Bromomethyl acetate, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC399180100

 View more versions of this product

Catalog No. AC399180100

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Powder
Formula Weight 152.97
Assay ≥94% (GC)
Quantity 10g
Color White to light beige
Refractive Index 1.4470 to 1.4540 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 1 Warning
Assay Percent Range 95%
CAS 590-97-6
Chemical Name or Material Bromomethyl acetate
Molecular Formula C3H5BrO2
MDL Number MFCD00000170
Linear Formula CH3CO2CH2Br
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Bromomethyl acetate
Assay ≥94% (GC)
Assay Percent Range 95%
Molecular Formula C3H5BrO2
Linear Formula CH3CO2CH2Br
Formula Weight 152.97
MDL Number MFCD00000170
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents