missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromomethyl acetate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 399180010

Catalog No. AC399180010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Bromomethyl acetate
1.5600g/mL
57°C
94% min. (GC)
C3H5BrO2
CH3CO2CH2Br
1g
bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
CC(=O)OCBr
152.98
152.97
590-97-6
130.0°C to 133.0°C (750.0mmHg)
Authentic
Glass bottle
1.4470 to 1.4540 (20°C, 589nm)
MFCD00000170
1.56
NHYXMAKLBXBVEO-UHFFFAOYSA-N
bromomethyl acetate
68536
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-696-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only