Please login to your online account to display your discounted pricing

Bromohydroquinone, 94%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC403040500

 View more versions of this product

Catalog No. AC403040500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Purity Grade Practical Grade
Formula Weight 189.01
Assay 39.7 to 42.7% (bromine, by combustion), Separat. Tech. GC ≥88%
Quantity 50g
Color Beige to brown
Melting Point 104° to 110°C
Assay Percent Range 94%
CAS 583-69-7
Chemical Name or Material Bromohydroquinone
Molecular Formula C6H5BrO2
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
MDL Number MFCD00041747
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Bromohydroquinone
CAS 583-69-7
Purity Grade Practical Grade
Assay 39.7 to 42.7% (bromine, by combustion), Separat. Tech. GC ≥88%
Assay Percent Range 94%
Molecular Formula C6H5BrO2
Formula Weight 189.01
MDL Number MFCD00041747
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents